accessibility menu, dialog, popup

UserName

ClCH3ON(S)(R)(S)OON(R)(S)(R)OCH3H

CAS RN 5786-96-9

IUPAC Name: hydrogen (1R,11S,18S,20R,21R,22S)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one chloride
Synonyms: hydrogen brucine chloride
Molecular Weight (g/mol): 430.93
Molecular Formula: C23H27ClN2O4
InChi Key: LOAGDNZFQUISCJ-SUJBTXFYSA-N
SMILES: [H+].[Cl-].COC1=CC2=C(C=C1OC)[C@@]13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results