missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromothymol Blue Sultone Form, ACS Reagent, Reag. Ph. Eur., Honeywell Fluka™
Indicator, ACS Reagent, Reag. Ph. Eur.
$104.18 - $4,824.14
Chemical Identifiers
| CAS | 76-59-5 |
|---|---|
| Molecular Formula | C27H28Br2O5S |
| Molecular Weight (g/mol) | 624.384 |
| MDL Number | MFCD00005872 |
| InChI Key | NUHCTOLBWMJMLX-UHFFFAOYSA-N |
| Synonym | 3â²,3â³-Dibromothymolsulfonphthalein |
| PubChem CID | 6450 |
| ChEBI | CHEBI:86155 |
| IUPAC Name | 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol |
| SMILES | CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 76-59-5 | |
| 624.384 | |
| NUHCTOLBWMJMLX-UHFFFAOYSA-N | |
| 6450 | |
| 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol |
| C27H28Br2O5S | |
| MFCD00005872 | |
| 3â²,3â³-Dibromothymolsulfonphthalein | |
| CHEBI:86155 | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br |
Specifications
| 76-59-5 | |
| MFCD00005872 | |
| 373934 | |
| NUHCTOLBWMJMLX-UHFFFAOYSA-N | |
| 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol | |
| 6450 | |
| 624.38g/mol | |
| Glass Bottle | |
| 5.8 to 7.4 | |
| 5 g | |
| Bromothymol Blue Sultone Form, Reag. Ph. Eur. |
| C27H28Br2O5S | |
| NONH for all modes of transport | |
| 3â²,3â³-Dibromothymolsulfonphthalein | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br | |
| 624.384 | |
| CHEBI:86155 | |
| ACS Reagent | |
| Complies for clarity of solution | |
| Violet | |
| Powder |
Safety and Handling
EINECSNumber : 200-971-2