missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromothymol Blue, ACS reagent

CAS: 76-59-5 | C27H28Br2O5S | 624.384 g/mol

$60.54 - $192.13

Chemical Identifiers

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.384
MDL Number MFCD00005872
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
Synonym Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB
PubChem CID 6450
ChEBI CHEBI:86155
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC403250050
SDS View Documents
Thermo Scientific Chemicals
403250050
5 g Glass bottle N/A
Only null left
 
AC403250100
SDS View Documents
Thermo Scientific Chemicals
403250100
10 g Glass bottle N/A
Only null left
 
AC403250250
SDS View Documents
Thermo Scientific Chemicals
403250250
25 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

76-59-5
624.384
NUHCTOLBWMJMLX-UHFFFAOYSA-N
6450
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
C27H28Br2O5S
MFCD00005872
Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB
CHEBI:86155
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br

Specifications

204.0°C
Passes Test
MFCD00005872
Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB
NUHCTOLBWMJMLX-UHFFFAOYSA-N
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
624.384
CHEBI:86155
ACS Reagent
from pH 6.0 (yellow) to pH 7.6 (blue)
5 g
Bromothymol Blue, Pure
76-59-5
C27H28Br2O5S
15, 1455
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, alkaline solution
Authentic
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
6450
624.38
Glass bottle
Brown/Pink to Purple
Crystalline Powder
Videos
SDS
Documents

Documents

RUO – Research Use Only