missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromothymol Blue, ACS reagent

CAS: 76-59-5 | C27H28Br2O5S | 624.384 g/mol

Supplier:  Thermo Scientific Chemicals 403250050

 View more versions of this product

Catalog No. AC403250050


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Bromothymol Blue
Pure
Passes Test
MFCD00005872
Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB
NUHCTOLBWMJMLX-UHFFFAOYSA-N
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
624.384
CHEBI:86155
ACS Reagent
from pH 6.0 (yellow) to pH 7.6 (blue)
5 g
204.0°C
76-59-5
C27H28Br2O5S
15, 1455
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, alkaline solution
Authentic
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
6450
624.38
Glass bottle
Brown/Pink to Purple
Crystalline Powder

Chemical Identifiers

76-59-5
624.384
NUHCTOLBWMJMLX-UHFFFAOYSA-N
6450
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
C27H28Br2O5S
MFCD00005872
Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB
CHEBI:86155
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only