missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromothymol Blue, pure, indicator

CAS: 76-59-5 | C27H28Br2O5S | 624.384 g/mol

Supplier:  Thermo Scientific Chemicals 151360250

 View more versions of this product

Catalog No. AC151360250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Bromothymol Blue
76-59-5
MFCD00005872
Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB
NUHCTOLBWMJMLX-UHFFFAOYSA-N
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
6450
624.38
Glass bottle
204.0°C
C27H28Br2O5S
15, 1455
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, alkaline solution
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
624.384
CHEBI:86155
Pure
25 g

Chemical Identifiers

76-59-5
624.384
NUHCTOLBWMJMLX-UHFFFAOYSA-N
6450
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
C27H28Br2O5S
MFCD00005872
Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB
CHEBI:86155
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-971-2

RTECSNumber : SJ7450000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only