missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromothymol Blue, pure, indicator

Catalog No. p-77459
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151360250 25 g Glass bottle
AC151361000 100 g Glass bottle
AC151365000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC151360250 Supplier Thermo Scientific Chemicals Supplier No. 151360250
Only null left

CAS: 76-59-5 | C27H28Br2O5S | 624.384 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.384
MDL Number MFCD00005872
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
Synonym Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB
PubChem CID 6450
ChEBI CHEBI:86155
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br

Specifications

Melting Point 204.0°C
Merck Index 15, 1455
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, alkaline solution
Formula Weight 624.38
Grade Pure
Packaging Glass bottle
Quantity 25 g
Chemical Name or Material Bromothymol Blue
EINECSNumber 200-971-2
RTECSNumber SJ7450000
TSCA TSCA

RUO – Research Use Only