missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue, 0.04% Aqueous, pH 6.0 to 7.6 Yellow to Blue, Certified, LabChem™

Supplier:  LabChem LC120502

Quantity 1 L
Packaging Poly Bottle

 View more versions of this product

Catalog No. LC120502


Only null left

Description

Description

Specifications

Chemical Identifiers

34722-90-2 , 7732-18-5
646.37
NMKFVGALBGZKGW-FKWCIMQXSA-M
sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
C27H27Br2NaO5S
MFCD00077263,MFCD00077263,MFCD00077263
102183223
[Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O

Specifications

Bromothymol Blue
34722-90-2 , 7732-18-5
C27H27Br2NaO5S
MFCD00077263,MFCD00077263,MFCD00077263
Passes Test
[Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O
646.37
646.37
Poly Bottle
Nitrogen oxides; Carbon monoxide; Carbon dioxide
Green
Liquid
0.04% Aqueous, pH 6.0 to 7.6 Yellow to Blue
99.96, 0.04
C27H27Br2O5S·Na
Soluble in water
NMKFVGALBGZKGW-FKWCIMQXSA-M
sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
102183223
Certified
1g/mL
1g/mL
1 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions