missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue, pure, 0.1% in water

CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol

$191.94 - $547.61

Specifications

Chemical Name or Material Bromophenol Blue
CAS 115-39-9 , 7732-18-5
Concentration about 0.1% (by UV)
Molecular Formula C19H10Br4O5S
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC403150250
View Documents Promotion Details
N/A 25 mL
Each for $191.94
Only null left
 
AC403151000
View Documents Promotion Details
N/A 100 mL
Each for $547.61
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Bromophenol Blue
about 0.1% (by UV)
3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
669.96
CHEBI:59424
Pure
1
115-39-9 , 7732-18-5
C19H10Br4O5S
UDSAIICHUKSCKT-UHFFFAOYSA-N
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1,lambda{6}-benzoxathiol-3-yl]phenol
8272
669.96
Glass bottle
1.0000g/mL
Videos
SDS
Documents

Documents

RUO – Research Use Only