missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue, pure, 0.1% in water

Catalog No. AC403150250
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC403150250 25 mL
AC403151000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC403150250 Supplier Thermo Scientific Chemicals Supplier No. 403150250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Specifications

Chemical Name or Material Bromophenol Blue
CAS 115-39-9, 7732-18-5
Concentration about 0.1% (by UV)
Molecular Formula C19H10Br4O5S
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1,lambda{6}-benzoxathiol-3-yl]phenol
Molecular Weight (g/mol) 669.96
PubChem CID 8272
ChEBI CHEBI:59424
Formula Weight 669.96
Grade Pure
Packaging Glass bottle
Specific Gravity 1
Density 1.0000g/mL
Quantity 25 mL
Show More Show Less

RUO – Research Use Only