missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue, ACS

CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol

$39.00 - $149.89

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,lambda {6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA3264103
View Documents Promotion Details
Thermo Scientific Chemicals
03264103
1 g
Each for $39.00
Only null left
 
AA3264106
View Documents Promotion Details
Thermo Scientific Chemicals
03264106
5 g
Each for $72.60
Only null left
 
AA3264114
View Documents Promotion Details
Thermo Scientific Chemicals
03264114
25 g
Each for $149.89
Only null left
 
Description

Description

Bromophenol blue is used as a tracking dye in electrophoresis, as a color marker in gel electrophoresis and polyacrylamide gel electrophoresis. As a biological stain, it plays an important role to stain proteins and nucleic acids. It acts an intermediate and an acid-base indicator in the pH range 3.0 to 4.6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

115-39-9
669.96
UDSAIICHUKSCKT-UHFFFAOYSA-N
CHEBI:59424
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
C19H10Br4O5S
MFCD00005875
8272
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,lambda {6}-benzoxathiol-3-yl]phenol

Specifications

270°C (decomposition)
C19H10Br4O5S
61698
Freely soluble in NaOH; Soluble in sodium hydroxide,alcohol,benzene,and acetic acid; Slightly soluble in water.
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
669.96
CHEBI:59424
ACS
Bromophenol Blue
115-39-9
MFCD00005875
141444
UDSAIICHUKSCKT-UHFFFAOYSA-N
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,lambda {6}-benzoxathiol-3-yl]phenol
8272
669.98
Crystalline
Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 204-086-2

RTECSNumber : SJ7453000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only