Learn More
Thermo Scientific Chemicals Bromophenol Blue, ACS
CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol
Supplier: Thermo Scientific Chemicals 03264106
| Quantity | 5 g |
|---|
Description
Bromophenol blue is used as a tracking dye in electrophoresis, as a color marker in gel electrophoresis and polyacrylamide gel electrophoresis. As a biological stain, it plays an important role to stain proteins and nucleic acids. It acts an intermediate and an acid-base indicator in the pH range 3.0 to 4.6.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 115-39-9 | |
| 669.96 | |
| UDSAIICHUKSCKT-UHFFFAOYSA-N | |
| CHEBI:59424 | |
| C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br |
| C19H10Br4O5S | |
| MFCD00005875 | |
| 8272 | |
| 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,lambda {6}-benzoxathiol-3-yl]phenol |
Specifications
| Bromophenol Blue | |
| 115-39-9 | |
| MFCD00005875 | |
| 141444 | |
| UDSAIICHUKSCKT-UHFFFAOYSA-N | |
| 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,lambda {6}-benzoxathiol-3-yl]phenol | |
| 8272 | |
| 669.98 | |
| Odorless | |
| Crystalline |
| 270°C (decomposition) | |
| C19H10Br4O5S | |
| 61698 | |
| Freely soluble in NaOH; Soluble in sodium hydroxide,alcohol,benzene,and acetic acid; Slightly soluble in water. | |
| C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br | |
| 669.96 | |
| CHEBI:59424 | |
| ACS | |
| 5 g |
Safety and Handling
H500
EINECSNumber : 204-086-2
RTECSNumber : SJ7453000
TSCA : Yes
Recommended Storage : Ambient temperatures