missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue, 0.1% Aqueous, pH 3.0 to 4.6 Yellow to Blue, Certified, LabChem™

Catalog No. p-5389087
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC120307 125 mL Poly Bottle
LC120301 500 mL Poly Bottle
LC120302 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC120307 Supplier LabChem Supplier No. LC120307
Only null left

Chemical Identifiers

CAS 62625-28-9 , 7732-18-5
Molecular Formula C19H9Br4NaO5S
Molecular Weight (g/mol) 691.942
InChI Key TWKDJMRHSLWRFI-UHFFFAOYSA-M
PubChem CID 3791690
IUPAC Name sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
SMILES C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]

Specifications

CAS Max % 99.9, 0.1
Linear Formula C19H9Br4O5S·Na
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 691.95
Grade Certified
Packaging Poly Bottle
Decomposition Information Hydrogen bromide; Carbon dioxide; Carbon monoxide; Nitrogen oxides
Color Purple/Red
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Bromophenol Blue, 0.1% Aqueous, pH 3.0 to 4.6 Yellow to Blue
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature