missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue, 0.04% Aqueous, pH 3.0 to 4.6 Yellow to Blue, Certified, LabChem™
$39.75 - $119.68
Chemical Identifiers
| CAS | 62625-28-9 |
|---|---|
| Molecular Formula | C19H9Br4NaO5S |
| Molecular Weight (g/mol) | 691.942 |
| InChI Key | TWKDJMRHSLWRFI-UHFFFAOYSA-M |
| PubChem CID | 3791690 |
| IUPAC Name | sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate |
| SMILES | C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+] |
Chemical Identifiers
| 62625-28-9 | |
| 691.942 | |
| 3791690 | |
| C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+] |
| C19H9Br4NaO5S | |
| TWKDJMRHSLWRFI-UHFFFAOYSA-M | |
| sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate |
Specifications
| 62625-28-9 , 7732-18-5 | |
| C19H9Br4NaO5S | |
| Soluble in water | |
| TWKDJMRHSLWRFI-UHFFFAOYSA-M | |
| sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate | |
| 3791690 | |
| Certified | |
| Hydrogen bromide; Carbon dioxide; Carbon monoxide; Nitrogen oxides | |
| 125 mL | |
| Bromophenol Blue, 0.04% Aqueous, pH 3.0 to 4.6 Yellow to Blue |
| 99.96,0.04 | |
| C19H9Br4O5S·Na | |
| Passes Test | |
| C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+] | |
| 691.942 | |
| 691.95 | |
| Poly Bottle | |
| Purple/Red | |
| Liquid |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature