missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromopentafluorobenzene, 99%
Description
It is used as pharmaceutical intermediate. It is a Grignard reagent (with CuBr in dioxane) can introduce pentafluorophenyl groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate. It is a Grignard reagent (with CuBr in dioxane) can introduce pentafluorophenyl groups.
Solubility
Insoluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
It is used as pharmaceutical intermediate. It is a Grignard reagent (with CuBr in dioxane) can introduce pentafluorophenyl groups.
Solubility
Insoluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
Specifications
Specifications
| Melting Point | ∼-31°C |
| Density | 1.981 |
| Boiling Point | 136°C to 137°C |
| Flash Point | 87°C (188°F) |
| Refractive Index | 1.45 |
| Quantity | 25 g |
| Beilstein | 1819390 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 246.97 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| EINECSNumber | 206-449-0 |
| RTECSNumber | CY9040552 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only