missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Bromomethyl)triphenylphosphonium Bromide 98.0+%, TCI America™
$98.79 - $277.75
Chemical Identifiers
| CAS | 1034-49-7 |
|---|---|
| Molecular Formula | C19H17Br2P |
| Molecular Weight (g/mol) | 436.13 |
| MDL Number | MFCD00011864 |
| InChI Key | YFTMLUSIDVFTKU-UHFFFAOYSA-M |
| Synonym | bromomethyl triphenylphosphonium bromide, bromomethyltriphenylphosphonium bromide, bromomethyl triphenylphosphoniumbromide, phosphonium, bromomethyl triphenyl-, bromide, bromomethyl triphenylphosphanium bromide, zlchem 395, bromomethyltriphenyl phosphonium bromide, bromomethyl triphenylphosphonium, bromomethyl triphenyl phosphanium bromide |
| PubChem CID | 2733422 |
| IUPAC Name | (bromomethyl)triphenylphosphanium bromide |
| SMILES | [Br-].BrC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 1034-49-7 | |
| 436.13 | |
| YFTMLUSIDVFTKU-UHFFFAOYSA-M | |
| 2733422 | |
| [Br-].BrC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C19H17Br2P | |
| MFCD00011864 | |
| bromomethyl triphenylphosphonium bromide, bromomethyltriphenylphosphonium bromide, bromomethyl triphenylphosphoniumbromide, phosphonium, bromomethyl triphenyl-, bromide, bromomethyl triphenylphosphanium bromide, zlchem 395, bromomethyltriphenyl phosphonium bromide, bromomethyl triphenylphosphonium, bromomethyl triphenyl phosphanium bromide | |
| (bromomethyl)triphenylphosphanium bromide |
Specifications
| 1034-49-7 | |
| White-Yellow | |
| MFCD00011864 | |
| bromomethyl triphenylphosphonium bromide, bromomethyltriphenylphosphonium bromide, bromomethyl triphenylphosphoniumbromide, phosphonium, bromomethyl triphenyl-, bromide, bromomethyl triphenylphosphanium bromide, zlchem 395, bromomethyltriphenyl phosphonium bromide, bromomethyl triphenylphosphonium, bromomethyl triphenyl phosphanium bromide | |
| [Br-].BrC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 436.13 | |
| 436.13 | |
| Crystalline Powder |
| 243°C | |
| C19H17Br2P | |
| 5 g | |
| YFTMLUSIDVFTKU-UHFFFAOYSA-M | |
| (bromomethyl)triphenylphosphanium bromide | |
| 2733422 | |
| ≥98.0% (T) | |
| (Bromomethyl)triphenylphosphonium Bromide |
Safety and Handling
TSCA : No