accessibility menu, dialog, popup

UserName

BrBrP

CAS RN 1034-49-7

IUPAC Name: (bromomethyl)triphenylphosphanium bromide
Synonyms: (bromomethyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 436.13
Molecular Formula: C19H17Br2P
InChi Key: YFTMLUSIDVFTKU-UHFFFAOYSA-M
SMILES: [Br-].BrC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results