missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)triphenylphosphonium bromide, 98%

Catalog No. AC321480050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC321480050 5 g Glass bottle
AC321480250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC321480050 Supplier Thermo Scientific Chemicals Supplier No. 321480050

CAS: 1034-49-7 | C19H17Br2P | 436.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 1034-49-7
Molecular Formula C19H17Br2P
Molecular Weight (g/mol) 436.13
MDL Number MFCD00011864
InChI Key YFTMLUSIDVFTKU-UHFFFAOYSA-M
Synonym bromomethyl triphenylphosphonium bromide, bromomethyltriphenylphosphonium bromide, bromomethyl triphenylphosphoniumbromide, phosphonium, bromomethyl triphenyl-, bromide, bromomethyl triphenylphosphanium bromide, zlchem 395, bromomethyltriphenyl phosphonium bromide, bromomethyl triphenylphosphonium, bromomethyl triphenyl phosphanium bromide
PubChem CID 2733422
IUPAC Name bromomethyl(triphenyl)phosphanium;bromide
SMILES [Br-].BrC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 230.0°C to 235.0°C
Color Beige to Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula BrCH2P(C6H5)3Br
Quantity 5 g
Formula Weight 436.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (Bromomethyl)triphenylphosphonium bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-857-2

RUO – Research Use Only