missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)cyclobutane 98.0+%, TCI America™

Catalog No. B36795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36795G 5 g
B367925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36795G Supplier TCI America Supplier No. B36795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17247-58-4
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.031
MDL Number MFCD00040960
InChI Key FLHFTXCMKFVKRP-UHFFFAOYSA-N
Synonym bromomethyl cyclobutane, cyclobutylmethyl bromide, cyclobutane, bromomethyl, sfphabikimup@, bromocyclobutylmethane, cyclobutylmethylbromide, bromomethyl-cyclobutane, cydobutyl-methylbromide, bromomethyl cyclobutan
PubChem CID 2734658
IUPAC Name bromomethylcyclobutane
SMILES C1CC(C1)CBr

Specifications

Color Yellow
Boiling Point 124°C
Quantity 5 g
UN Number 1993
Formula Weight 149.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Bromomethyl)cyclobutane