accessibility menu, dialog, popup

UserName

Br

CAS RN 17247-58-4

IUPAC Name: (bromomethyl)cyclobutane
Synonyms: (bromomethyl)cyclobutane
Molecular Weight (g/mol): 149.03
Molecular Formula: C5H9Br
InChi Key: FLHFTXCMKFVKRP-UHFFFAOYSA-N
SMILES: BrCC1CCC1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)