missing translation for 'onlineSavingsMsg'
Learn More

Bromoethane 99.0+%, TCI America™

Catalog No. p-7118545
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
B058825G 25 g
B0588500G 500 g
2 options

Catalog No. B058825G

Supplier: TCI America B058825G

Only null left

Chemical Identifiers

CAS 74-96-4
Molecular Formula C2H5Br
Molecular Weight (g/mol) 108.966
MDL Number MFCD00000232
InChI Key RDHPKYGYEGBMSE-UHFFFAOYSA-N
Synonym ethyl bromide, ethane, bromo, 1-bromoethane, monobromoethane, bromic ether, hydrobromic ether, bromure d'ethyle, ethylbromide, etylu bromek, halon 2001
PubChem CID 6332
IUPAC Name bromoethane
SMILES CCBr

Specifications

Color Yellow
Boiling Point 38°C
Quantity 25 g
UN Number 1891
Formula Weight 108.97
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromoethane
EINECSNumber (9)-0518
RTECSNumber KH6475000
TSCA Yes
Recommended Storage Refrigerator