accessibility menu, dialog, popup

UserName

H3CBr

CAS RN 74-96-4

IUPAC Name: bromoethane
Synonyms: bromoethane
Molecular Weight (g/mol): 108.97
Molecular Formula: C2H5Br
InChi Key: RDHPKYGYEGBMSE-UHFFFAOYSA-N
SMILES: CCBr
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)