missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclopropane 98.0+%, TCI America™

Catalog No. p-7124772
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Catalog No. Quantity
C10675G 5 g
C106725G 25 g
C1067250G 250 g
3 options

Catalog No. C10675G

Supplier: TCI America C10675G

Only null left

Chemical Identifiers

CAS 4333-56-6
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.977
MDL Number MFCD00001271
InChI Key LKXYJYDRLBPHRS-UHFFFAOYSA-N
Synonym cyclopropyl bromide, cyclopropane, bromo, cyclopropylbromide, bromo-cyclopropan, bromo-cyclopropane, cyclopropylbrornide, bromanylcyclopropane, pubchem14417, bromocyclopropane, acmc-1aii5
PubChem CID 78037
IUPAC Name bromocyclopropane
SMILES C1CC1Br

Specifications

Color Yellow
Boiling Point 69°C
Quantity 5 g
UN Number 1993
Formula Weight 120.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromocyclopropane