missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclopropane, 99%

Catalog No. p-7022291
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1499609 10 g
AAA1499618 50 g
AAA1499630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1499609 Supplier Thermo Scientific Chemicals Supplier No. A1499609

CAS: 4333-56-6 | C3H5Br | 120.977 g/mol

Bromo Cyclopropane is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It is also used in the production of cyclopropyl boric acid. It is used as an intermediate in organic synethesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromo Cyclopropane is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It is also used in the production of cyclopropyl boric acid. It is used as an intermediate in organic synethesis.

Solubility
It is soluble in chloroform, and methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents, strong bases.

Chemical Identifiers

CAS 4333-56-6
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.977
MDL Number MFCD00001271
InChI Key LKXYJYDRLBPHRS-UHFFFAOYSA-N
Synonym cyclopropyl bromide, cyclopropane, bromo, cyclopropylbromide, bromo-cyclopropan, bromo-cyclopropane, cyclopropylbrornide, bromanylcyclopropane, pubchem14417, bromocyclopropane, acmc-1aii5
PubChem CID 78037
IUPAC Name bromocyclopropane
SMILES C1CC1Br

Specifications

Density 1.515
Boiling Point 68°C to 70°C
Flash Point −6°C (21°F)
Odor Recognizable
Refractive Index 1.46
Quantity 10 g
UN Number UN1993
Beilstein 1900287
Solubility Information It is soluble in chloroform,and methanol. Insoluble in water.
Formula Weight 120.98
Percent Purity 99%
Chemical Name or Material Bromocyclopropane
Show More Show Less
Hazard Category H224-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 224-375-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only