missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclobutane, 95%

CAS: 4399-47-7 | C4H7Br | 135.004 g/mol

Supplier:  Thermo Scientific Chemicals A1281706

 View more versions of this product

Catalog No. AAA1281706


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bromocyclobutane
1.431
22°C (71°F)
1.4795
5 g
1900415
cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h
C1CC(C1)Br
135.004
135.01
4399-47-7
107°C to 108°C
C4H7Br
MFCD00001317
UN1993
Light sensitive
KXVUSQIDCZRUKF-UHFFFAOYSA-N
bromocyclobutane
78110
95%

Chemical Identifiers

4399-47-7
135.004
KXVUSQIDCZRUKF-UHFFFAOYSA-N
78110
C1CC(C1)Br
C4H7Br
MFCD00001317
cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h
bromocyclobutane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H225-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 224-530-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only