missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocyclobutane, 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.431 |
| Boiling Point | 107°C to 108°C |
| Flash Point | 22°C (71°F) |
| Refractive Index | 1.4795 |
| Quantity | 1 g |
| UN Number | UN1993 |
| Beilstein | 1900415 |
| Sensitivity | Light sensitive |
| Formula Weight | 135.01 |
| Percent Purity | 95% |
| Show More |
Safety and Handling
| Hazard Category | H225-H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 224-530-9 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only