missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Purple sodium salt

CAS: 62625-30-3 | C21H15Br2NaO5S | 562.204 g/mol

Supplier:  Thermo Scientific Chemicals A1908218

 View more versions of this product

Catalog No. AAA1908218


Only null left

Description

Description

Bromocresol purple sodium salt is used to isolate acid-producing bacteria from sauerkraut juice, and lactic bacteria is scalped by morphology and share, then morphological, physiological and biochemical interactions. It can be used as a fluorescent stain to detect yeast cells with plasma membrane damage for dead cell counts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocresol purple sodium salt is used to isolate acid-producing bacteria from sauerkraut juice, and lactic bacteria is scalped by morphology and share, then morphological, physiological and biochemical interactions. It can be used as a fluorescent stain to detect yeast cells with plasma membrane damage for dead cell counts.

Solubility
Soluble in water.

Notes
Transition interval: pH 5.2 (yellow) to pH 6.8 (purple)
Specifications

Specifications

Bromocresol Purple sodium salt
62625-30-3
MFCD00148896
Soluble in water.
CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]
562.204
562.22
∼255°C (decomposition)
C21H15Br2NaO5S
Bromocresol Purple, water soluble
KVSJYIXUWNOWBD-UGAWPWHASA-M
sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
23665568
50 g

Chemical Identifiers

62625-30-3
562.204
KVSJYIXUWNOWBD-UGAWPWHASA-M
23665568
CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]
C21H15Br2NaO5S
MFCD00148896
Bromocresol Purple, water soluble
sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 263-655-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only