missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Purple sodium salt

Catalog No. p-7024435
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1908209 10 g
AAA1908218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1908209 Supplier Thermo Scientific Chemicals Supplier No. A1908209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62625-30-3 | C21H15Br2NaO5S | 562.204 g/mol

Bromocresol purple sodium salt is used to isolate acid-producing bacteria from sauerkraut juice, and lactic bacteria is scalped by morphology and share, then morphological, physiological and biochemical interactions. It can be used as a fluorescent stain to detect yeast cells with plasma membrane damage for dead cell counts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocresol purple sodium salt is used to isolate acid-producing bacteria from sauerkraut juice, and lactic bacteria is scalped by morphology and share, then morphological, physiological and biochemical interactions. It can be used as a fluorescent stain to detect yeast cells with plasma membrane damage for dead cell counts.

Solubility
Soluble in water.

Notes
Transition interval: pH 5.2 (yellow) to pH 6.8 (purple)

Chemical Identifiers

CAS 62625-30-3
Molecular Formula C21H15Br2NaO5S
Molecular Weight (g/mol) 562.204
MDL Number MFCD00148896
InChI Key KVSJYIXUWNOWBD-UGAWPWHASA-M
Synonym Bromocresol Purple, water soluble
PubChem CID 23665568
IUPAC Name sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
SMILES CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]

Specifications

Melting Point ∼255°C (decomposition)
Solubility Information Soluble in water.
Quantity 10 g
Formula Weight 562.22
Chemical Name or Material Bromocresol Purple sodium salt
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 263-655-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only