missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Green, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$472.65 - $472.65
Chemical Identifiers
| CAS | 76-60-8 |
|---|---|
| Molecular Formula | C21H14Br4O5S |
| Molecular Weight (g/mol) | 698.014 |
| InChI Key | FRPHFZCDPYBUAU-UHFFFAOYSA-N |
| PubChem CID | 6451 |
| IUPAC Name | 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
| SMILES | CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br |
Chemical Identifiers
| 76-60-8 | |
| 698.014 | |
| 6451 | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br |
| C21H14Br4O5S | |
| FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
| 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
Specifications
| 76-60-8 | |
| FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
| 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol | |
| 6451 | |
| BAKER ANALYZED™ ACS Reagent | |
| 5 g |
| C21H14Br4O5S | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br | |
| 698.014 | |
| 698.02 | |
| Wide Mouth Amber Glass Bottle | |
| Bromocresol Green |