Learn More
Thermo Scientific Chemicals Bromocresol Green, ACS reagent
CAS: 76-60-8 | C21H14Br4O5S | 698.014 g/mol
$48.73 - $155.54
Chemical Identifiers
| CAS | 76-60-8 |
|---|---|
| Molecular Formula | C21H14Br4O5S |
| Molecular Weight (g/mol) | 698.014 |
| MDL Number | MFCD00005874 |
| InChI Key | FRPHFZCDPYBUAU-UHFFFAOYSA-N |
| Synonym | 3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5' |
| PubChem CID | 6451 |
| IUPAC Name | 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
| SMILES | CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC402930010
|
Thermo Scientific Chemicals
402930010 |
1 g | Glass bottle |
Each for $48.73
|
|
||||
|
AC402930050
|
Thermo Scientific Chemicals
402930050 |
5 g | Glass bottle |
Each for $91.31
|
|
||||
|
AC402930100
|
Thermo Scientific Chemicals
402930100 |
10 g | Glass bottle |
Each for $155.54
Case of 6 Each for $0.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 76-60-8 | |
| 698.014 | |
| FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
| 6451 | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br |
| C21H14Br4O5S | |
| MFCD00005874 | |
| 3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5' | |
| 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
Specifications
| 225.0°C | |
| Passes Test | |
| MFCD00005874 | |
| 3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5' | |
| FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br | |
| 698.014 | |
| 698.01 | |
| Glass bottle | |
| Beige to Brown | |
| Powder |
| 76-60-8 | |
| C21H14Br4O5S | |
| 15, 1393 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in benzene, ethanol (30g/L), very soluble in diethyl ether and ethylacetate | |
| Authentic | |
| 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol | |
| 6451 | |
| ACS Reagent | |
| from pH 3.8 (yellow) to pH 5.4 (blue) | |
| 1 g | |
| Bromocresol Green |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 200-972-8
RTECSNumber : SJ7456000
TSCA : TSCA
RUO – Research Use Only