missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green, ACS reagent

CAS: 76-60-8 | C21H14Br4O5S | 698.014 g/mol

Supplier:  Thermo Scientific Chemicals 402930100

 View more versions of this product

Catalog No. AC402930100



Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Bromocresol Green
Passes Test
MFCD00005874
3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5'
Authentic
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
6451
ACS Reagent
from pH 3.8 (yellow) to pH 5.4 (blue)
10 g
76-60-8
C21H14Br4O5S
15, 1393
FRPHFZCDPYBUAU-UHFFFAOYSA-N
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br
698.014
698.01
Glass bottle
Beige to Brown
Powder

Chemical Identifiers

76-60-8
698.014
FRPHFZCDPYBUAU-UHFFFAOYSA-N
6451
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br
C21H14Br4O5S
MFCD00005874
3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5'
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-972-8

RTECSNumber : SJ7456000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only