missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Green, ACS Reagent (Sultone) not water soluble, Reagents
Bromocresol Green, ACS Reagent (Sultone) not water soluble, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$87.14 - $1156.89
Chemical Identifiers
| CAS | 76-60-8 |
|---|---|
| Molecular Formula | C21H14Br4O5S |
| Molecular Weight (g/mol) | 698.01 |
| InChI Key | FRPHFZCDPYBUAU-UHFFFAOYSA-N |
| Synonym | Bromocresol blue, Bromcresol Green, Bromcresol Blue |
| IUPAC Name | 3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione |
| SMILES | CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-336-783
|
Labchem Inc
C6105500100C2 |
100 g |
Each for $1,156.89
|
|
|||||
|
01-336-782
|
Labchem Inc
C610550010C1 |
10 g |
Each for $330.39
|
|
|||||
|
01-336-781
|
Labchem Inc
C61055001C2 |
1 g |
Each for $87.14
|
|
|||||
|
01-336-780
|
Labchem Inc
C61055005C2 |
5 g |
Each for $198.75
|
|
|||||
Chemical Identifiers
| 76-60-8 | |
| 698.01 | |
| Bromocresol blue, Bromcresol Green, Bromcresol Blue | |
| CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C |
| C21H14Br4O5S | |
| FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
| 3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione |
Specifications
| 76-60-8 | |
| Bromocresol blue, Bromcresol Green, Bromcresol Blue | |
| CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C | |
| 698.01 | |
| Glass Bottle | |
| Brown to Yellow | |
| Solid |
| C21H14Br4O5S | |
| FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
| 3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione | |
| ACS | |
| 0.79 | |
| 100 g | |
| Bromocresol Green, ACS Reagent (Sultone) not water soluble |