missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green, ACS Reagent (Sultone) not water soluble, Reagents

Bromocresol Green, ACS Reagent (Sultone) not water soluble, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc C610550010C1

 View more versions of this product

Catalog No. 01-336-782


Only null left

Description

Description

Specifications

Specifications

Bromocresol Green, ACS Reagent (Sultone) not water soluble
C21H14Br4O5S
FRPHFZCDPYBUAU-UHFFFAOYSA-N
3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
ACS
0.79
10 g
76-60-8
Bromocresol blue, Bromcresol Green, Bromcresol Blue
CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C
698.01
Glass Bottle
Brown to Yellow
Solid

Chemical Identifiers

76-60-8
698.01
Bromocresol blue, Bromcresol Green, Bromcresol Blue
CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C
C21H14Br4O5S
FRPHFZCDPYBUAU-UHFFFAOYSA-N
3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions