missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bromoacetonitrile 97.0+%, TCI America™
Supplier: TCI America B105825G
Specifications
Bromoacetonitrile | |
Yellow | |
C2H2BrN | |
25 g | |
bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrile | |
BrCC#N | |
119.95 | |
119.95 | |
Liquid |
590-17-0 | |
150°C | |
MFCD00001884 | |
3276 | |
REXUYBKPWIPONM-UHFFFAOYSA-N | |
2-bromoacetonitrile | |
11534 | |
≥97.0% (GC) |
Chemical Identifiers
590-17-0 | |
119.95 | |
REXUYBKPWIPONM-UHFFFAOYSA-N | |
11534 | |
BrCC#N |
C2H2BrN | |
MFCD00001884 | |
bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrile | |
2-bromoacetonitrile |
Safety and Handling
RTECSNumber : AL7970000
TSCA : No