missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetonitrile 97.0+%, TCI America™

Catalog No. p-7118826
Change view
Click to view available options
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
B105825G 25 g
B1058100G 100 g
B1058500G 500 g
3 options

Catalog No. B105825G

Supplier: TCI America B105825G

Only null left

Chemical Identifiers

CAS 590-17-0
Molecular Formula C2H2BrN
Molecular Weight (g/mol) 119.95
MDL Number MFCD00001884
InChI Key REXUYBKPWIPONM-UHFFFAOYSA-N
Synonym bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrile
PubChem CID 11534
IUPAC Name 2-bromoacetonitrile
SMILES BrCC#N

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 g
UN Number 3276
Formula Weight 119.95
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromoacetonitrile
RTECSNumber AL7970000
TSCA No