missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-p-xylene, 98%

Catalog No. p-3735454
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107570050 5 g Glass bottle
AC107570250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107570050 Supplier Thermo Scientific Chemicals Supplier No. 107570050
Only null left

CAS: 104-81-4 | C8H9Br | 185.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-81-4
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000181
InChI Key WZRKSPFYXUXINF-UHFFFAOYSA-N
Synonym 4-methylbenzyl bromide, 1-bromomethyl-4-methylbenzene, p-xylyl bromide, p-methylbenzyl bromide, alpha-bromo-p-xylene, benzene, 1-bromomethyl-4-methyl, 4-bromomethyl toluene, p-bromomethyl toluene, alpha-bromo-p-xylol, 1-bromomethyl-4-methyl-benzene
PubChem CID 7721
IUPAC Name 1-(bromomethyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)CBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33.0°C to 37.0°C
Color White to Yellow
Boiling Point 218.0°C to 220.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2Br
Quantity 5 g
Beilstein 05, 385
Merck Index 15, 10288
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in chloroform and ether
Formula Weight 185.06
Percent Purity 98%
Physical Form Crystalline Mass
Chemical Name or Material α-Bromo-p-xylene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-240-6

RUO – Research Use Only