accessibility menu, dialog, popup

UserName

CAS RN 104-81-4

IUPAC Name: 1-(bromomethyl)-4-methylbenzene
Synonyms: 1-(bromomethyl)-4-methylbenzene
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: WZRKSPFYXUXINF-UHFFFAOYSA-N
SMILES: CC1=CC=C(CBr)C=C1
Boiling Point 
  • (3)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (6)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)