missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-2,3,4,5,6-pentafluorotoluene, 97%

CAS: 1765-40-8 | C7H2BrF5 | 260.98 g/mol

$119.82 - $194.29

Chemical Identifiers

CAS 1765-40-8
MDL Number MFCD00000299
InChI Key XDEPVFFKOVDUNO-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide
PubChem CID 74484
IUPAC Name 1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC146330050
SDS View Documents
Thermo Scientific Chemicals
146330050
5 g Glass bottle
Each for $119.82
Only null left
 
AC146330100
SDS View Documents
Thermo Scientific Chemicals
146330100
10 g Glass bottle
Each for $194.29
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1765-40-8
XDEPVFFKOVDUNO-UHFFFAOYSA-N
74484
C(C1=C(C(=C(C(=C1F)F)F)F)F)Br
MFCD00000299
2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide
1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene

Specifications

1765-40-8
100.0
Colorless to Yellow
174.0°C to 175.0°C
Authentic
Glass bottle
1.4700 to 1.4720
MFCD00000299
1.72
XDEPVFFKOVDUNO-UHFFFAOYSA-N
1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene
74484
97%
α-Bromo-2,3,4,5,6-pentafluorotoluene
96.0
19.0°C
1.7200g/mL
82°C
97%
C7H2BrF5
C6F5CH2Br
5 g
2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide
C(C1=C(C(=C(C(=C1F)F)F)F)F)Br
260.989
260.98
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 217-182-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only