missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-2,3,4,5,6-pentafluorotoluene, 97%

CAS: 1765-40-8 | C7H2BrF5 | 260.98 g/mol

Supplier:  Thermo Scientific Chemicals 146330100

 View more versions of this product

Catalog No. AC146330100


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

α-Bromo-2,3,4,5,6-pentafluorotoluene
96.0
19.0°C
1.7200g/mL
82°C
97%
C7H2BrF5
C6F5CH2Br
10 g
2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide
XDEPVFFKOVDUNO-UHFFFAOYSA-N
1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene
74484
97%
1765-40-8
100.0
Colorless to Yellow
174.0°C to 175.0°C
Authentic
Glass bottle
1.4700 to 1.4720
MFCD00000299
1.72
Solubility in water: insoluble
C(C1=C(C(=C(C(=C1F)F)F)F)F)Br
260.98
260.98
Liquid

Chemical Identifiers

1765-40-8
XDEPVFFKOVDUNO-UHFFFAOYSA-N
74484
C(C1=C(C(=C(C(=C1F)F)F)F)F)Br
MFCD00000299
2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide
1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 217-182-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only