missing translation for 'onlineSavingsMsg'
Learn More

Bromine - 1,4-Dioxane Complex 95.0+%, TCI America™

Supplier:  TCI America B271925G

 View more versions of this product

Catalog No. B271925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bromine - 1,4-Dioxane Complex
25 g
C4H8BrO2
p-dioxane, compd. with bromine 1:1
C1COCCO1.[Br]
168.01
247.91
15481-39-7
Crystal-Powder at 20°C
1325
GSQACUNMFGRAKY-UHFFFAOYSA-N
bromine;1,4-dioxane
57348859
≥95.0% (T)

Chemical Identifiers

15481-39-7
168.01
p-dioxane, compd. with bromine 1:1
bromine;1,4-dioxane
C4H8BrO2
GSQACUNMFGRAKY-UHFFFAOYSA-N
57348859
C1COCCO1.[Br]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions