accessibility menu, dialog, popup

UserName

BrBrOO

CAS RN 15481-39-7

IUPAC Name: 1,4-dioxane; dibromine
Synonyms: Brom; dioxane
Molecular Weight (g/mol): 247.91
Molecular Formula: C4H8Br2O2
InChi Key: DTLTUUJDCNTTSN-UHFFFAOYSA-N
SMILES: BrBr.C1COCCO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results