missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromide Standard for Ion Chromatography, 1000μg/mL Br-, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$330.03 - $648.25
Chemical Identifiers
| CAS | 7647-15-6 |
|---|---|
| Molecular Formula | BrNa |
| Molecular Weight (g/mol) | 102.89 |
| MDL Number | MFCD00003475 |
| InChI Key | JHJLBTNAGRQEKS-UHFFFAOYSA-M |
| Synonym | sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a |
| PubChem CID | 253881 |
| ChEBI | CHEBI:63004 |
| IUPAC Name | sodium bromide |
| SMILES | [Na+].[Br-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
ASBR9-2Y
|
SPEX CertiPrep Group LLC
ASBR92Y |
125 mL | Triple-leached LDPE bottle |
Each for $330.03
|
|
||||
|
AS-BR9-2X
|
SPEX CertiPrep Group LLC
ASBR92X |
500 mL | Triple-leached LDPE bottle |
Each for $648.25
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Br- | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7647-15-6 | |
| 102.89 | |
| JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
| 253881 | |
| sodium bromide |
| BrNa | |
| MFCD00003475 | |
| sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
| CHEBI:63004 | |
| [Na+].[Br-] |
Specifications
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,99.9 | |
| MFCD00003475 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Standard Solution | |
| JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
| Water | |
| 102.89 | |
| 253881 | |
| 1000μg/mL Br- |
| 7647-15-6,7732-18-5 | |
| 0.1,99.9 | |
| BrNa | |
| Ion Chromatography | |
| sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
| Certified Reference Material | |
| [Na+].[Br-] | |
| sodium bromide | |
| 125 mL | |
| CHEBI:63004 | |
| Bromide Standard |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only