missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromide Standard for Ion Chromatography, 1000μg/mL Br-, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC ASBR92X
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Br- | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Bromide Standard | |
| 7647-15-6,7732-18-5 | |
| 0.1,99.9 | |
| BrNa | |
| Ion Chromatography | |
| sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
| Certified Reference Material | |
| [Na+].[Br-] | |
| sodium bromide | |
| 500 mL | |
| CHEBI:63004 |
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,99.9 | |
| MFCD00003475 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Standard Solution | |
| JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
| Water | |
| 102.89 | |
| 253881 | |
| 1000μg/mL Br- |
Chemical Identifiers
| 7647-15-6 | |
| 102.89 | |
| JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
| 253881 | |
| sodium bromide |
| BrNa | |
| MFCD00003475 | |
| sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
| CHEBI:63004 | |
| [Na+].[Br-] |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only