missing translation for 'onlineSavingsMsg'
Learn More

Bromide Standard, 1000 ppm Br-, Fisherbrand

Catalog No. ACCU0822500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACCU0822500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACCU0822500 Supplier Ricca Chemical Company Supplier No. ACCU0822500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7647-15-6 , 7732-18-5
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
PubChem CID 253881
ChEBI CHEBI:63004
IUPAC Name sodium bromide
SMILES [Na+].[Br-]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
Solubility Miscible with water
Solution Type Calibration Standard
Type Single-component Standard
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Bromide, 1000ppm Br
Show More Show Less