missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromate-Bromide Solution, 0.100 N (N/10), Ricca Chemical
Supplier: Ricca Chemical Company RI11701
Specifications
| Bromate-Bromide Solution | |
| 7732-18-5,7758-01-2,7758-01-2 | |
| 100.0,1.01,0.28 | |
| 167 g/mol | |
| Amber Glass Bottle | |
| Liquid |
| Colorless | |
| 97.0,0.97,0.27 | |
| dihydrogen oxide, dihydrogen monoxide | |
| Laboratory | |
| 4 L | |
| 0.100Normal |