missing translation for 'onlineSavingsMsg'
Learn More

Bromate-Bromide Solution, 0.100 N (N/10), Ricca Chemical

Catalog No. p-3251382
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
117016 500 mL Amber Glass Bottle
117032 1 L Amber Glass Bottle
11701 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 117016 Supplier Ricca Chemical Company Supplier No. 117016
Only null left

Specifications

Chemical Name or Material Bromate-Bromide Solution
Color Colorless
CAS 7758-01-2, 7758-02-3, 7758-01-2
CAS Min % 97.0, 0.97, 0.27
CAS Max % 100.0, 1.01, 0.28
Molecular Formula BrKO3
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
SMILES [O-]Br(=O)=O.[K+]
IUPAC Name potassium;bromate
Molecular Weight (g/mol) 167.00
PubChem CID 23673461
ChEBI CHEBI:38211
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.100Normal
Show More Show Less