missing translation for 'onlineSavingsMsg'
Learn More

Brivudine, 98%

CAS: 69304-47-8 | C11H13BrN2O5 | 333.14 g/mol

$147.11 - $1,065.47

Chemical Identifiers

CAS 69304-47-8
Molecular Formula C11H13BrN2O5
Molecular Weight (g/mol) 333.14
MDL Number MFCD00058585
InChI Key ODZBBRURCPAEIQ-PIXDULNESA-N
Synonym brivudine, bvdu, brivudin, helpin, e-5-2-bromovinyl-2'-deoxyuridine, bromovinyldeoxyuridine, zostex, brivudine inn, brivudinum inn-latin, brivudina inn-spanish
PubChem CID 446727
IUPAC Name 5-[(E)-2-bromoethenyl]-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(\C=C\Br)C(=O)NC1=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH27853MF
View Documents Promotion Details
N/A 50 mg
Each for $147.11
Only null left
 
AAH27853MD
View Documents Promotion Details
N/A 250 mg
Each for $386.49
Only null left
 
AAH2785303
View Documents Promotion Details
N/A 1 g
Each for $1,065.47
Only null left
 
Description

Description

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in water and methanol.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

69304-47-8
333.14
ODZBBRURCPAEIQ-PIXDULNESA-N
446727
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(\C=C\Br)C(=O)NC1=O
C11H13BrN2O5
MFCD00058585
brivudine, bvdu, brivudin, helpin, e-5-2-bromovinyl-2'-deoxyuridine, bromovinyldeoxyuridine, zostex, brivudine inn, brivudinum inn-latin, brivudina inn-spanish
5-[(E)-2-bromoethenyl]-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione

Specifications

69304-47-8
C11H13BrN2O5
brivudine, bvdu, brivudin, helpin, e-5-2-bromovinyl-2'-deoxyuridine, bromovinyldeoxyuridine, zostex, brivudine inn, brivudinum inn-latin, brivudina inn-spanish
ODZBBRURCPAEIQ-PIXDULNESA-N
+10° (c=1 in Methanol)
333.14
333.14
(E)-5-(2-Bromovinyl)-2'-deoxyuridine
∽165°C (decomposition)
MFCD00058585
Soluble in water and methanol.
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(\C=C\Br)C(=O)NC1=O
5-[(E)-2-bromoethenyl]-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
446727
98%
Videos
Safety and Handling

Safety and Handling

RTECSNumber : YU7355000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only