missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brivudine, 98%
CAS: 69304-47-8 | C11H13BrN2O5 | 333.14 g/mol
Supplier: Thermo Scientific Chemicals H27853MD
| Quantity | 250 mg |
|---|
Description
It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate.
Solubility
Soluble in water and methanol.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 69304-47-8 | |
| 333.14 | |
| ODZBBRURCPAEIQ-PIXDULNESA-N | |
| 446727 | |
| OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(\C=C\Br)C(=O)NC1=O |
| C11H13BrN2O5 | |
| MFCD00058585 | |
| brivudine, bvdu, brivudin, helpin, e-5-2-bromovinyl-2'-deoxyuridine, bromovinyldeoxyuridine, zostex, brivudine inn, brivudinum inn-latin, brivudina inn-spanish | |
| 5-[(E)-2-bromoethenyl]-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
Specifications
| (E)-5-(2-Bromovinyl)-2'-deoxyuridine | |
| ∽165°C (decomposition) | |
| 250 mg | |
| brivudine, bvdu, brivudin, helpin, e-5-2-bromovinyl-2'-deoxyuridine, bromovinyldeoxyuridine, zostex, brivudine inn, brivudinum inn-latin, brivudina inn-spanish | |
| ODZBBRURCPAEIQ-PIXDULNESA-N | |
| +10° (c=1 in Methanol) | |
| 333.14 | |
| 333.14 |
| 69304-47-8 | |
| C11H13BrN2O5 | |
| MFCD00058585 | |
| Soluble in water and methanol. | |
| OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(\C=C\Br)C(=O)NC1=O | |
| 5-[(E)-2-bromoethenyl]-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione | |
| 446727 | |
| 98% |
Safety and Handling
RTECSNumber : YU7355000
TSCA : No
Recommended Storage : Keep cold