missing translation for 'onlineSavingsMsg'
Learn More

Brivudine, 98%

Catalog No. AAH27853MF
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27853MF 50 mg
AAH27853MD 250 mg
AAH2785303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH27853MF Supplier Thermo Scientific Chemicals Supplier No. H27853MF
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69304-47-8 | C11H13BrN2O5 | 333.14 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in water and methanol.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 69304-47-8
Molecular Formula C11H13BrN2O5
Molecular Weight (g/mol) 333.14
MDL Number MFCD00058585
InChI Key ODZBBRURCPAEIQ-PIXDULNESA-N
Synonym brivudine, bvdu, brivudin, helpin, e-5-2-bromovinyl-2'-deoxyuridine, bromovinyldeoxyuridine, zostex, brivudine inn, brivudinum inn-latin, brivudina inn-spanish
PubChem CID 446727
IUPAC Name 5-[(E)-2-bromoethenyl]-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(\C=C\Br)C(=O)NC1=O

Specifications

Melting Point ∽165°C (decomposition)
Quantity 50 mg
Solubility Information Soluble in water and methanol.
Optical Rotation +10° (c=1 in Methanol)
Formula Weight 333.14
Percent Purity 98%
Chemical Name or Material (E)-5-(2-Bromovinyl)-2'-deoxyuridine
RTECSNumber YU7355000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only