missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Brivaracetam
Description
Brivaracetam is an antiepileptic compound that is a ligand for synaptic vesicle glycoprotein 2A (SV2A; Kd = 50 nM for human SV2A in vitro). Brivaracetam displays selectivity for SV2A at 10 μM, against a panel of 55 receptors, enzymes, ion channels and transporters. In mice, Brivaracetam protects against clonic audiogenic seizures.
Specifications
Specifications
| Chemical Name or Material | (αS,4R)-α-Ethyl-2-oxo-4-propyl-1-pyrrolidineacetamide |
| CAS | 357336-20-0 |
| Quantity | 10 mg |
| Target | Translocation, Exo/Endocytosis Products |
| Molecular Formula | C11H20N2O2 |
| Purity | 0.98 |