missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brij™-58 Surfact-Amps™ Detergent Solution

Catalog No. PI28336
Change view
Click to view available options
Quantity:
6 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI28336 6 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI28336 Supplier Thermo Scientific™ Supplier No. LSG28336

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purified Brij-58 detergent that is stabilized and supplied as a 10% solution in 10mL glass ampules for protein research and molecular biology.

Thermo Scientific™ Surfact-Amps 58 Detergent Solution is highly-purified Brij™-58 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.

This is an easy-to-use 10% (w/v) solution of purified Brij™-58 detergent for use in routine and high-demand protein research methods and molecular biology techniques. Brij-58 is a nonionic polyoxyethylene surfactant that is most frequently used in HPLC applications.

Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipet and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Highlights:

Brij-58 – a nonionic detergent for use as a surfactant in HPLC

Accurate – precise 10% detergent solution in ultrapure water

Easy-to-use – solution is simple to dispense and dilute for use

Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls

Stable – packed in glass ampules under inert nitrogen gas

Detergent Properties:

  • Molecular Weight: 1120
  • Detergent Class: Nonionic
  • Aggregation Number: 70
  • Micelle Molecular Weight: 82,000
  • Critical Micelle Concentration (CMC): 0.077mM (0.0086%, w/v)
  • Cloud Point: >100°C
  • Dialyzable: No

Specifications

Product Type Detergent
Content And Storage Store at Room Temperature
Form Liquid
Product Line Brij, Surfact-Amps
Reagent Type Detergent Solution
Quantity 6 x 10 mL

For Research Use Only. Not for use in diagnostic procedures.